Gene Transcripts

Gene Symbol
Slc43a1
Entrez Gene ID
311168
Gene Name
solute carrier family 43 (amino acid system L transporter), member 1
Gene Aliases
Chromosome Location
Chr. 3: 72238796 - 72265023 on Build Rnor_6.0
UniGene ID
Rn.35465
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107742.1 NP_001101212.1 870
GenBank mRNA BC161926.1 - 831

View Full Product Details