Gene Transcripts

Gene Symbol
Paxip1
Entrez Gene ID
311944
Gene Name
PAX interacting (with transcription-activation domain) protein 1
Gene Aliases
Chromosome Location
Chr. 4: 3959640 - 4008326 on Build Rnor_6.0
UniGene ID
Rn.136961
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107844.1 NP_001101314.1 9, 10 2037

View Full Product Details