Gene Transcripts

Gene Symbol
Mier1
Entrez Gene ID
313418
Gene Name
mesoderm induction early response 1, transcriptional regulator
Gene Aliases
RGD1562337
Chromosome Location
Chr. 5: 122642063 - 122693642 on Build Rnor_6.0
UniGene ID
Rn.36542
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001131012.1 NP_001124484.1 13 1339
XM_006238479.1 XP_006238541.1 14 1698
XM_006238483.2 XP_006238545.1 15 1455
XM_006238484.2 XP_006238546.1 15 1565
XM_008763874.1 XP_008762096.1 13 4981
XM_008763875.1 XP_008762097.1 14 4422
XM_008763876.1 XP_008762098.1 14 4253
XM_008763877.1 XP_008762099.1 14 4897
XM_008763878.1 XP_008762100.1 14 2421
XM_008763879.1 XP_008762101.1 14 2418
XM_008763880.1 XP_008762102.1 11 1238
XM_008763881.1 XP_008762103.1 14 1615
GenBank mRNA BC168252.1 - 13 1339

View Full Product Details