Gene Transcripts

Gene Symbol
ARSD
Entrez Gene ID
414
Gene Name
arylsulfatase D
Gene Aliases
ASD
Chromosome Location
Chr. X: 2903970 - 2929375 on Build GRCh38
UniGene ID
Hs.528631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001669.3 NP_001660.2 2 214
NM_009589.3 NP_033667.2 2 214
XM_005274514.2 XP_005274571.1 2 184
XM_005274515.2 XP_005274572.1 2 184
GenBank mRNA AF160499.1 - 2 114
AK314479.1 - 2 178
BC020229.1 - 142
BC080179.1 - 2 117
DA869167.1 - 2 214
X83572.1 - 2 190

View Full Product Details