Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006241942.2 | XP_006242004.1 | 4 | 1254 | |
XM_006241943.2 | XP_006242005.1 | 5 | 1136 | ||
XM_006241944.2 | XP_006242006.1 | 4 | 971 | ||
XM_006241945.1 | XP_006242007.1 | 4 | 707 | ||
XM_006241946.2 | XP_006242008.1 | 4 | 951 | ||
XM_006241947.2 | XP_006242009.1 | 5 | 1027 | ||
XM_006241948.2 | XP_006242010.1 | 5 | 999 | ||
XM_008765644.1 | XP_008763866.1 | 4 | 727 | ||
XM_008765645.1 | XP_008763867.1 | 5 | 979 | ||
XM_008765646.1 | XP_008763868.1 | 2 | 567 | ||
XM_235465.8 | XP_235465.6 | 5 | 1157 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_003750381.3 | XP_003750429.1 | 1158 | ||
XM_006241953.2 | XP_006242015.1 | 1255 | |||
XM_006241954.2 | XP_006242016.1 | 1137 | |||
XM_006241955.2 | XP_006242017.1 | 4 | 972 | ||
XM_006241956.1 | XP_006242018.1 | 707 | |||
XM_006241957.2 | XP_006242019.1 | 4 | 952 | ||
XM_006241958.2 | XP_006242020.1 | 5 | 1028 | ||
XM_006241959.2 | XP_006242021.1 | 5 | 1000 | ||
XM_008765799.1 | XP_008764021.1 | 727 | |||
XM_008765800.1 | XP_008764022.1 | 5 | 980 | ||
XM_008765801.1 | XP_008764023.1 | 2 | 567 |