Gene Transcripts

Gene Symbol
Wdr13
Entrez Gene ID
317370
Gene Name
WD repeat domain 13
Gene Aliases
RGD1560982
Chromosome Location
Chr. X: 15112665 - 15123825 on Build Rnor_6.0
UniGene ID
Rn.107767
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108247.2 NP_001101717.1 3 428
XM_006256719.2 XP_006256781.1 4 969
XM_006256720.2 XP_006256782.1 4 1226
XM_008773075.1 XP_008771297.1 4 1197
GenBank mRNA AF452730.1 - 4 1176
BC167040.1 - 3 428 7135266

View Full Product Details