Gene Transcripts
- Gene Symbol
- Myh4
- Entrez Gene ID
- 360543
- Gene Name
- myosin, heavy chain 4, skeletal muscle
- Gene Aliases
- Chromosome Location
- Chr. 10: 53778456 - 53801605 on Build Rnor_6.0
- UniGene ID
- Rn.224464
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019325.1 |
NP_062198.1
|
19 |
2313 |
|
GenBank mRNA |
BC113948.1 |
- |
19 |
2313 |
|
View Full Product Details