Gene Transcripts

Gene Symbol
B3GALT2
Entrez Gene ID
8707
Gene Name
beta-1,3-galactosyltransferase 2
Gene Aliases
BETA3GALT2, GLCT2, beta3Gal-T2
Chromosome Location
Chr. 1: 193178730 - 193186613 on Build GRCh38
UniGene ID
Hs.518834
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003783.3 NP_003774.1 2 1009
GenBank mRNA AF288390.1 - 2 989
AK314116.1 - 2 727
BC022507.1 - 2 1009 4792498
Y15060.1 - 1 294

View Full Product Details