Gene Transcripts

Gene Symbol
Ulk1
Entrez Gene ID
360827
Gene Name
unc-51 like autophagy activating kinase 1
Gene Aliases
Chromosome Location
Chr. 12: 51908142 - 51934704 on Build Rnor_6.0
UniGene ID
Rn.24509
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108341.1 NP_001101811.1 1 100
XM_006249565.2 XP_006249627.1 1 406

View Full Product Details