Gene Transcripts
- Gene Symbol
- Ms4a6a
- Entrez Gene ID
- 361735
- Gene Name
- membrane-spanning 4-domains, subfamily A, member 6A
- Gene Aliases
- Ms4a11
- Chromosome Location
- Chr. 1: 227919284 - 227932637 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_342028.7 |
XP_342029.5
|
7 |
853 |
|
View Full Product Details