Gene Transcripts

Gene Symbol
Aifm2
Entrez Gene ID
361843
Gene Name
apoptosis-inducing factor, mitochondrion-associated 2
Gene Aliases
Amid, Prg3
Chromosome Location
Chr. 20: 31327697 - 31353548 on Build Rnor_6.0
UniGene ID
Rn.1357
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001139483.1 NP_001132955.1 3 359
XM_006256452.2 XP_006256514.1 4 473
XM_006256453.2 XP_006256515.1 4 473
XM_006256454.2 XP_006256516.1 4 416
XM_006256456.2 XP_006256518.1 4 443
XM_006256459.2 XP_006256521.1 4 473
XM_008772888.1 XP_008771110.1 4 473
XM_008772889.1 XP_008771111.1 4 431
XM_008772890.1 XP_008771112.1 4 418
GenBank mRNA AW521424.1 - 3 262

View Full Product Details