Gene Transcripts

Gene Symbol
CACNA1I
Entrez Gene ID
8911
Gene Name
calcium voltage-gated channel subunit alpha1 I
Gene Aliases
Cav3.3, ca(v)3.3
Chromosome Location
Chr. 22: 39570753 - 39689737 on Build GRCh38
UniGene ID
Hs.125116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003406.1 NP_001003406.1 4 499
NM_021096.3 NP_066919.2 4 499
GenBank mRNA AF129133.1 - 4 499
AF142567.1 - 4 499
AF211189.1 - 4 499
AF393329.1 - 4 499

View Full Product Details