Gene Transcripts

Gene Symbol
Ndufa1
Entrez Gene ID
363441
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1
Gene Aliases
RGD1560955
Chromosome Location
Chr. X: 124513143 - 124516791 on Build Rnor_6.0
UniGene ID
Rn.3640
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108813.2 NP_001102283.2 2 228
GenBank mRNA BC168213.1 - 2 204
FM054992.1 - 2 213
FQ214979.1 - 2 189

Gene Symbol
NEWGENE_1560955
Entrez Gene ID
103694507
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1, 7.5kDa
Gene Aliases
Ndufa1
Chromosome Location
Chr. X: 123803103 - 123806851 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773489.1 XP_008771711.1 2 319

View Full Product Details