Gene Transcripts

Gene Symbol
Vom1r76
Entrez Gene ID
494240
Gene Name
vomeronasal 1 receptor 76
Gene Aliases
V1rc27
Chromosome Location
Chr. 4: 87852749 - 87853648 on Build Rnor_6.0
UniGene ID
Rn.129664
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009520.1 NP_001009520.1 1 345
GenBank mRNA AY510327.1 - 1 345

View Full Product Details