Gene Transcripts

Gene Symbol
Rnase17
Entrez Gene ID
497195
Gene Name
ribonuclease 17
Gene Aliases
R17
Chromosome Location
Chr. 15: 28204043 - 28204929 on Build Rnor_6.0
UniGene ID
Rn.142436
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012232.1 NP_001012232.1 1 184
XM_008770561.1 XP_008768783.1 2 262

View Full Product Details