Gene Transcripts

Gene Symbol
Arl8a
Entrez Gene ID
498234
Gene Name
ADP-ribosylation factor-like 8A
Gene Aliases
RGD1565940
Chromosome Location
Chr. 13: 51972974 - 51981976 on Build Rnor_6.0
UniGene ID
Rn.138927
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109071.1 NP_001102541.1 4, 5 537

View Full Product Details