Gene Transcripts

Gene Symbol
Arl13a
Entrez Gene ID
501617
Gene Name
ADP-ribosylation factor-like 13A
Gene Aliases
Chromosome Location
Chr. X: 105010968 - 105039080 on Build Rnor_6.0
UniGene ID
Rn.123458
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024366.1 NP_001019537.1 7, 8 1023
XM_008773430.1 XP_008771652.1 7, 8 1544
GenBank mRNA BC079325.1 - 7, 8 1023

View Full Product Details