Gene Transcripts

Gene Symbol
Aarsd1
Entrez Gene ID
619440
Gene Name
alanyl-tRNA synthetase domain containing 1
Gene Aliases
Chromosome Location
Chr. 10: 89317974 - 89331078 on Build Rnor_6.0
UniGene ID
Rn.153971
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034109.1 NP_001029281.1 4 359
GenBank mRNA BC083790.1 - 4 359
FQ213285.1 - 4 360

View Full Product Details