Gene Transcripts

Gene Symbol
Ripply1
Entrez Gene ID
680702
Gene Name
ripply transcriptional repressor 1
Gene Aliases
Chromosome Location
Chr. X: 111099420 - 111102539 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773499.1 XP_008771721.1 4 463

View Full Product Details