Gene Transcripts

Gene Symbol
Prss41
Entrez Gene ID
681033
Gene Name
protease, serine, 41
Gene Aliases
RGD1304769, Tessp1
Chromosome Location
Chr. 10: 13179781 - 13187461 on Build Rnor_6.0
UniGene ID
Rn.220927
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135087.1 NP_001128559.1 6 693

View Full Product Details