Gene Transcripts

Gene Symbol
Snrpe
Entrez Gene ID
100360682
Gene Name
small nuclear ribonucleoprotein polypeptide E
Gene Aliases
Chromosome Location
Chr. 13: 50252707 - 50258951 on Build Rnor_6.0
UniGene ID
Rn.145709
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271237.1 NP_001258166.1 5 355
GenBank mRNA FQ211122.1 - 5 355

View Full Product Details