Gene Transcripts

Gene Symbol
Lmo3
Entrez Gene ID
497798
Gene Name
LIM domain only 3
Gene Aliases
RGD1561357
Chromosome Location
Chr. 4: 172942027 - 173005630 on Build Rnor_6.0
UniGene ID
Rn.127746
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749863.3 XP_003749911.1 2 675
XM_006237585.2 XP_006237647.1 2 659
XM_006237588.2 XP_006237650.1 3 667
XM_006237590.2 XP_006237652.1 2 273
XM_006237591.2 XP_006237653.1 2 453
XM_008763453.1 XP_008761675.1 2 603
GenBank mRNA FQ213730.1 - 1 418

View Full Product Details