Gene Transcripts

Gene Symbol
Bloc1s3
Entrez Gene ID
680476
Gene Name
biogenesis of lysosomal organelles complex-1, subunit 3
Gene Aliases
Chromosome Location
Chr. 1: 80414766 - 80417352 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228499.2 XP_006228561.1 2 1010

View Full Product Details