Gene Transcripts

Gene Symbol
Morn3
Entrez Gene ID
687615
Gene Name
MORN repeat containing 3
Gene Aliases
Chromosome Location
Chr. 12: 38961329 - 38976475 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249367.2 XP_006249429.1 4 818
XM_008769278.1 XP_008767500.1 4 793
XM_008769279.1 XP_008767501.1 5 848

View Full Product Details