Gene Transcripts

Gene Symbol
PSMD6
Entrez Gene ID
9861
Gene Name
proteasome 26S subunit, non-ATPase 6
Gene Aliases
Rpn7, S10, SGA-113M, p42A, p44S10
Chromosome Location
Chr. 3: 64010549 - 64024010 on Build GRCh38
UniGene ID
Hs.152536
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271779.1 NP_001258708.1 6 1117
NM_001271780.1 NP_001258709.1 5 832
NM_001271781.1 NP_001258710.1 5 707
NM_014814.2 NP_055629.1 5 958
XM_005265619.1 XP_005265676.1 5 1054
XM_011534288.1 XP_011532590.1 6 1013
XM_017007569.1 XP_016863058.1 6 860
GenBank mRNA AF215935.1 - 5 824
AF530062.1 - 5 771
AK095059.1 - 4 1540
AK290205.1 - 5 812
AL529900.3 - 5 693
AY359879.1 - 5 804
AY568086.1 - 5 796
BC000630.1 - 5 788 3344973
BC000904.2 - 5 809 2900971
BC012369.1 - 5 806 3858094
BX647742.1 - 6 984
D14663.1 - 5 796

View Full Product Details