Gene Transcripts

Gene Symbol
LY6E
Entrez Gene ID
4061
Gene Name
lymphocyte antigen 6 complex, locus E
Gene Aliases
RIG-E, RIGE, SCA-2, SCA2, TSA-1
Chromosome Location
Chr. 8: 143018485 - 143022410 on Build GRCh38
UniGene ID
Hs.521903
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127213.1 NP_001120685.1 4 393
NM_002346.2 NP_002337.1 4 398
GenBank mRNA AF085875.1 - 2 134
AK129615.1 - 4 355
AK223074.1 - 4 377
AK311983.1 - 4 354
BC003392.1 - 4 302
BC119708.1 - 3 279 40117169
BC119709.1 - 3 279 40117170
DB458093.1 - 4 398
U42376.1 - 4 333
U56145.1 - 3 239
Z68179.1 - 4 333

View Full Product Details