Gene Transcripts

Gene Symbol
LACTBL1
Entrez Gene ID
646262
Gene Name
lactamase beta like 1
Gene Aliases
Chromosome Location
Chr. 1: 22953043 - 22972316 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289974.1 NP_001276903.1 6 1627

View Full Product Details