Gene Transcripts

Gene Symbol
EBI3
Entrez Gene ID
10148
Gene Name
Epstein-Barr virus induced 3
Gene Aliases
IL-27B, IL27B
Chromosome Location
Chr. 19: 4229196 - 4237539 on Build GRCh38
UniGene ID
Hs.501452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005755.2 NP_005746.2 3 282
XM_011527619.2 XP_011525921.1 4 531
GenBank mRNA BC015364.1 - 3 327
BC046112.1 - 3 281 5748376
L08187.1 - 3 287

View Full Product Details