Gene Transcripts

Gene Symbol
ATP9B
Entrez Gene ID
374868
Gene Name
ATPase phospholipid transporting 9B (putative)
Gene Aliases
ATPASEP, ATPIIB, HUSSY-20, NEO1L, hMMR1
Chromosome Location
Chr. 18: 79069275 - 79378391 on Build GRCh38
UniGene ID
Hs.465475
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK128847.1 - 1 1973

Gene Symbol
LOC100996274
Entrez Gene ID
100996274
Gene Name
uncharacterized LOC100996274
Gene Aliases
Chromosome Location
Chr. 18: 79144752 - 79149853 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011526289.2 XP_011524591.1 1 1976

View Full Product Details