Gene Transcripts

Gene Symbol
LINC01548
Entrez Gene ID
728409
Gene Name
long intergenic non-protein coding RNA 1548
Gene Aliases
C21orf54
Chromosome Location
Chr. 21: 33165470 - 33170236 on Build GRCh38
UniGene ID
Hs.125234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024102.1 4 408
GenBank mRNA BC105115.1 - 4 408
BC105117.1 - 4 408
BX116511.1 - 4 361

View Full Product Details