Gene Transcripts

Gene Symbol
AKAP8
Entrez Gene ID
10270
Gene Name
A-kinase anchoring protein 8
Gene Aliases
AKAP 95, AKAP-8, AKAP-95, AKAP95
Chromosome Location
Chr. 19: 15353521 - 15379815 on Build GRCh38
UniGene ID
Hs.594496
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005858.3 NP_005849.1 5 582
XM_011527624.2 XP_011525926.1 5 675
XM_017026141.1 XP_016881630.1 5 727
GenBank mRNA AK303640.1 - 2 210
AK308251.1 - 3 319
BC037270.2 - 5 572 4837309
DR005936.1 - 5 585
Y11997.1 - 5 573

View Full Product Details