Gene Transcripts

Gene Symbol
Acbd3
Entrez Gene ID
170760
Gene Name
acyl-Coenzyme A binding domain containing 3
Gene Aliases
60kDa, 8430407O11Rik, D1Ertd10e, GCP60, GOLPH1, Gocap1, Pap7
Chromosome Location
Chr. 1: 180726043 - 180754204 on Build GRCm38
UniGene ID
Mm.272981
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133225.3 NP_573488.2 8 1513
GenBank mRNA AF022770.3 - 8 1514
AK030371.1 - 8 1514
AK168799.1 - 8 1487
BC060602.1 - 8 1468 5706182

View Full Product Details