Gene Transcripts

Gene Symbol
NMUR1
Entrez Gene ID
10316
Gene Name
neuromedin U receptor 1
Gene Aliases
(FM-3), FM-3, FM3, GPC-R, GPR66, NMU1R
Chromosome Location
Chr. 2: 231519903 - 231530471 on Build GRCh38
UniGene ID
Hs.471619
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006056.4 NP_006047.3 3 2310
XM_011510487.2 XP_011508789.1 3 2298
GenBank mRNA BC036543.1 - 3 2338

View Full Product Details