Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195518.1 | NP_001182447.1 | 3 | 386 | |
NM_006077.3 | NP_006068.2 | 3 | 386 | ||
XM_005269383.2 | XP_005269440.1 | 3 | 290 | ||
XM_011539119.1 | XP_011537421.1 | 3 | 290 | ||
GenBank mRNA | AK023318.1 | - | 3 | 305 | |
AK225298.1 | - | 3 | 290 | ||
AK225695.1 | - | 3 | 274 | ||
AL117423.1 | - | 3 | 277 | ||
BC004190.2 | - | 3 | 298 | 3009677 | |
BC004216.2 | - | 3 | 298 | 3009677 | |
BC016641.1 | - | 3 | 261 | 3919114 | |
DB030067.1 | - | 3 | 336 |