Gene Transcripts
- Gene Symbol
- TACC3
- Entrez Gene ID
- 10460
- Gene Name
- transforming acidic coiled-coil containing protein 3
- Gene Aliases
- ERIC-1, ERIC1
- Chromosome Location
- Chr. 4: 1721383 - 1745178 on Build GRCh38
- UniGene ID
- Hs.104019
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006342.2 |
NP_006333.1
|
8, 9 |
1911 |
|
XM_005247929.2 |
XP_005247986.1
|
8, 9 |
1909 |
|
XM_005247930.2 |
XP_005247987.1
|
7, 8 |
938 |
|
XM_011513386.2 |
XP_011511688.1
|
8, 9 |
1909 |
|
XM_017007653.1 |
XP_016863142.1
|
8, 9 |
1963 |
|
GenBank mRNA |
AF093543.1 |
- |
8, 9 |
1864 |
|
AF479620.1 |
- |
7, 8 |
777 |
|
AJ243997.1 |
- |
8, 9 |
1856 |
|
AK302463.1 |
- |
8, 9 |
1882 |
|
AK304579.1 |
- |
7, 8 |
1640 |
|
AK310660.1 |
- |
7, 8 |
1634 |
|
BC106071.1 |
- |
8, 9 |
1872 |
6148176 |
BC111771.1 |
- |
8, 9 |
1824 |
40034847 |
View Full Product Details