Gene Transcripts

Gene Symbol
Ptx4
Entrez Gene ID
68509
Gene Name
pentraxin 4
Gene Aliases
1110018H23Rik
Chromosome Location
Chr. 17: 25120697 - 25125286 on Build GRCm38
UniGene ID
Mm.328855
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163416.1 NP_001156888.1 2 640
NM_026747.1 NP_081023.1 2 730
GenBank mRNA BC107380.2 - 2 623
BC107381.2 - 2 713 40057088
BC118508.1 - 2 713 40106397

View Full Product Details