Gene Transcripts

Gene Symbol
Naa40
Entrez Gene ID
70999
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit, homolog (S. cerevisiae)
Gene Aliases
4931433E08Rik, AU023197, Nat11
Chromosome Location
Chr. 19: 7225668 - 7241222 on Build GRCm38
UniGene ID
Mm.329605
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027643.1 NP_081919.1 4 503
GenBank mRNA AK016504.1 - 4 464
AK033510.1 - 4 325
AK033896.1 - 4 334
AK035418.1 - 3 1221
AK162625.1 - 4 337
AK170548.1 - 4 329
AK187772.1 - 1 77
AK193575.1 - 1 78
AK213481.1 - 3 230
AK216197.1 - 3 231
BC020020.1 - 4 354 3156924

View Full Product Details