Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006421.4 | NP_006412.2 | 36 | 5369 | |
XM_005251134.4 | XP_005251191.1 | 37 | 5348 | ||
XM_005251135.3 | XP_005251192.1 | 36 | 5601 | ||
XM_005251136.3 | XP_005251193.1 | 36 | 5619 | ||
XM_011517442.2 | XP_011515744.1 | 37 | 5655 | ||
GenBank mRNA | AB209324.1 | - | 26 | 3414 | |
AF084520.1 | - | 36 | 4979 | ||
AF111162.1 | - | 36 | 5120 | ||
AK001788.1 | - | 9 | 1013 | ||
AK022557.1 | - | 12 | 1468 | ||
AK025637.1 | - | 17 | 1968 | ||
AK127799.1 | - | 3 | 2567 | ||
AL117446.1 | - | 2 | 72 |