Gene Transcripts

Gene Symbol
BRDT
Entrez Gene ID
676
Gene Name
bromodomain testis associated
Gene Aliases
BRD6, CT9
Chromosome Location
Chr. 1: 91949371 - 92014428 on Build GRCh38
UniGene ID
Hs.482520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242805.2 NP_001229734.2 19 3147
NM_001242806.2 NP_001229735.2 18 3090
NM_001242807.2 NP_001229736.2 17 2746
NM_001242808.2 NP_001229737.2 17 2940
NM_001242810.2 NP_001229739.2 18 3434
NM_001726.4 NP_001717.3 18 2873
NM_207189.3 NP_997072.2 18 3078
XM_006710853.3 XP_006710916.1 19 3663
XM_006710854.3 XP_006710917.1 20 3508
XM_006710855.3 XP_006710918.1 20 3513
XM_006710856.3 XP_006710919.1 20 3487
XM_006710857.3 XP_006710920.1 20 3477
XM_011542032.2 XP_011540334.1 20 2997
XM_011542033.2 XP_011540335.1 20 2971
XM_011542034.2 XP_011540336.1 20 2987
XM_011542035.2 XP_011540337.1 20 2961
XM_011542036.2 XP_011540338.1 18 2876
GenBank mRNA AK302758.1 - 17 2682
AK303008.1 - 18 2838
AK316442.1 - 17 2745
AY338951.1 - 19 3147
BX648135.1 - 18 3169

View Full Product Details