Gene Transcripts

Gene Symbol
Akip1
Entrez Gene ID
361624
Gene Name
A kinase (PRKA) interacting protein 1
Gene Aliases
RGD1306959
Chromosome Location
Chr. 1: 174330630 - 174339494 on Build Rnor_6.0
UniGene ID
Rn.106344
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108497.1 NP_001101967.1 2, 3 519
XM_006229987.1 XP_006230049.1 3, 4 408
XM_006229988.2 XP_006230050.1 3, 4 339
XM_006229989.1 XP_006230051.1 3, 4 378
XM_006229990.1 XP_006230052.1 3, 4 408

View Full Product Details