Gene Transcripts

Gene Symbol
ARPC4
Entrez Gene ID
10093
Gene Name
actin related protein 2/3 complex subunit 4
Gene Aliases
ARC20, P20-ARC
Chromosome Location
Chr. 3: 9792495 - 9807105 on Build GRCh38
UniGene ID
Hs.323342
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024959.2 NP_001020130.1 6 1266
NM_001024960.2 NP_001020131.1 6 1121
NM_001198780.1 NP_001185709.1 6 1120
NM_005718.4 NP_005709.1 6 1632
GenBank mRNA AK074205.1 - 1 1735
BC012596.2 - 5 1051

View Full Product Details