Gene Transcripts

Gene Symbol
CPM
Entrez Gene ID
1368
Gene Name
carboxypeptidase M
Gene Aliases
Chromosome Location
Chr. 12: 68842200 - 68963240 on Build GRCh38
UniGene ID
Hs.654387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005502.2 NP_001005502.1 6 766
NM_001874.4 NP_001865.1 6 798
NM_198320.3 NP_938079.1 6 784
GenBank mRNA AF368463.1 - 6 756
AK130146.1 - 3 374
AK313180.1 - 6 756
BC022276.1 - 6 756 4768771
DA833423.1 - 3 325
J04970.2 - 6 798

View Full Product Details