Gene Transcripts

Gene Symbol
PRKX
Entrez Gene ID
5613
Gene Name
protein kinase, X-linked
Gene Aliases
PKX1
Chromosome Location
Chr. X: 3604343 - 3713634 on Build GRCh38
UniGene ID
Hs.390788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005044.4 NP_005035.1 7 1270
GenBank mRNA BC041073.1 - 7 1073
X85545.1 - 7 1255

View Full Product Details