Gene Transcripts

Gene Symbol
ANKRD26
Entrez Gene ID
22852
Gene Name
ankyrin repeat domain 26
Gene Aliases
THC2, bA145E8.1
Chromosome Location
Chr. 10: 26938195 - 27100498 on Build GRCh38
UniGene ID
Hs.361041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256053.1 NP_001242982.1 21 2451
NM_014915.2 NP_055730.2 21 2454
XM_006717423.2 XP_006717486.1 22 3540
XM_006717425.3 XP_006717488.1 22 3540
XM_011519416.2 XP_011517718.1 22 3540
XM_017015928.1 XP_016871417.1 22 3540
XM_017015929.1 XP_016871418.1 23 3528
XM_017015930.1 XP_016871419.1 22 3540
XM_017015931.1 XP_016871420.1 22 3540
XM_017015932.1 XP_016871421.1 22 3540
XM_017015933.1 XP_016871422.1 22 3540
GenBank mRNA AB028997.1 - 21 2429
AK128577.1 - 6 1014
BC130453.1 - 21 2395 40146484

View Full Product Details