Gene Transcripts

Gene Symbol
ADRM1
Entrez Gene ID
11047
Gene Name
adhesion regulating molecule 1
Gene Aliases
ARM-1, ARM1, GP110
Chromosome Location
Chr. 20: 62302056 - 62308862 on Build GRCh38
UniGene ID
Hs.90107
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281437.1 NP_001268366.1 7 1010
NM_001281438.1 NP_001268367.1 7 1014
NM_007002.3 NP_008933.2 8 1131
NM_175573.2 NP_783163.1 8 1127
XM_005260257.1 XP_005260314.1 9 1218
XM_011528503.1 XP_011526805.1 8 1966
XM_017027602.1 XP_016883091.1 8 1987
XM_017027603.1 XP_016883092.1 9 1239
XM_017027604.1 XP_016883093.1 8 1983
XM_017027605.1 XP_016883094.1 8 1987
XM_017027606.1 XP_016883095.1 7 1870
XM_017027607.1 XP_016883096.1 7 1866
XM_017027608.1 XP_016883097.1 6 1656
XM_017027609.1 XP_016883098.1 6 1635
XM_017027610.1 XP_016883099.1 6 1635
GenBank mRNA AK054985.1 - 1928
AK307459.1 - 1061
BC010733.1 - 5 643
BC017245.2 - 8 1027 4890786
CR749213.1 - 2 4543
D64154.1 - 8 1022

View Full Product Details