Gene Transcripts

Gene Symbol
OR2M4
Entrez Gene ID
26245
Gene Name
olfactory receptor family 2 subfamily M member 4
Gene Aliases
HSHTPCRX18, HTPCRX18, OR1-55, OST710, TPCR100
Chromosome Location
Chr. 1: 248238929 - 248239864 on Build GRCh38
UniGene ID
Hs.381309
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017504.1 NP_059974.1 1 583
GenBank mRNA X64992.1 - 1 209
X89666.1 - 1 209

View Full Product Details