Gene Transcripts

Gene Symbol
SPANXN2
Entrez Gene ID
494119
Gene Name
SPANX family member N2
Gene Aliases
CT11.7, SPANX-N2
Chromosome Location
Chr. X: 143712035 - 143720668 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA DQ336114.1 - 2 238

View Full Product Details