Gene Transcripts

Gene Symbol
C1rb
Entrez Gene ID
667277
Gene Name
complement component 1, r subcomponent B
Gene Aliases
Gm8551, mC1rB
Chromosome Location
Chr. 6: 124570430 - 124581044 on Build GRCm38
UniGene ID
Mm.481484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113356.1 NP_001106827.1 1828
GenBank mRNA AF459018.1 - 1828

View Full Product Details