Gene Transcripts
- Gene Symbol
- Ptms
- Entrez Gene ID
- 69202
- Gene Name
- parathymosin
- Gene Aliases
- 2610009E16Rik
- Chromosome Location
- Chr. 6: 124913675 - 124917946 on Build GRCm38
- UniGene ID
- Mm.142187
- Species
- Mus musculus
- Gene Symbol
- Osbpl7
- Entrez Gene ID
- 71240
- Gene Name
- oxysterol binding protein-like 7
- Gene Aliases
- 4933437E18Rik
- Chromosome Location
- Chr. 11: 97050344 - 97068904 on Build GRCm38
- UniGene ID
- Mm.190450
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC085242.1 |
- |
|
1112 |
|
View Full Product Details