Gene Transcripts

Gene Symbol
Rgs11
Entrez Gene ID
50782
Gene Name
regulator of G-protein signaling 11
Gene Aliases
C78048
Chromosome Location
Chr. 17: 26202962 - 26211324 on Build GRCm38
UniGene ID
Mm.109840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081069.1 NP_001074538.1 12 799
GenBank mRNA AK039314.1 - 11 1421
AK161551.1 - 8 858
BC049910.1 - 10 1503

View Full Product Details